Assay Details
| Gene Symbol: Vamp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299326.1 | NP_001286255.1 | 4 - 5 | 882 | 66 | |
| NM_136680.4 | NP_610524.1 | 4 - 5 | 824 | 66 | ||
| GenBank mRNA | AY071695.1 | - | 4 - 5 | 836 | 66 | |
More Information
Set Membership: |
|

Vamp7
Genomic Map