Assay Details
| Gene Symbol: Pdrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136694.4 | NP_610538.1 | 1 - 2 | 295 | 103 | |
| GenBank mRNA | BT023615.1 | - | 1 - 2 | 261 | 103 | |
More Information
Set Membership: |
|

Pdrg1
Genomic Map