Assay Details
| Gene Symbol: eIF3j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299331.1 | NP_001286260.1 | 4 - 5 | 669 | 75 | |
| NM_136697.3 | NP_610541.1 | 4 - 5 | 804 | 75 | ||
| GenBank mRNA | AY051457.1 | - | 4 - 5 | 649 | 75 | |
More Information
Set Membership: |
|

eIF3j
Genomic Map