Assay Details
| Gene Symbol: PIG-N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136705.3 | NP_610549.1 | 1 - 2 | 812 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIG-N| Gene Symbol: PIG-N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136705.3 | NP_610549.1 | 1 - 2 | 812 | 89 | |
Set Membership: |
|