Assay Details
| Gene Symbol: RpLP0-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136710.4 | NP_610554.1 | 2 - 3 | 375 | 67 | |
| GenBank mRNA | AY061517.1 | - | 2 - 3 | 331 | 67 | |
More Information
Set Membership: |
|

RpLP0-like
Genomic Map