Assay Details
| Gene Symbol: SLO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103788.3 | NP_001097258.2 | 13 - 14 | 2236 | 114 | |
| NM_001103789.3 | NP_001097259.2 | 13 - 14 | 2236 | 114 | ||
| NM_001201971.2 | NP_001188900.1 | 13 - 14 | 2532 | 114 | ||
| NM_001201972.3 | NP_001188901.1 | 13 - 14 | 2749 | 114 | ||
| NM_001259313.2 | NP_001246242.1 | 13 - 14 | 2749 | 114 | ||
| NM_001273932.1 | NP_001260861.1 | 13 - 14 | 2749 | 114 | ||
| NM_001273933.1 | NP_001260862.1 | 12 - 13 | 1739 | 114 | ||
| NM_001273934.1 | NP_001260863.1 | 13 - 14 | 2228 | 114 | ||
| NM_001273935.1 | NP_001260864.1 | 13 - 14 | 2749 | 114 | ||
More Information
Set Membership: |
|

SLO2
Genomic Map