Assay Details
| Gene Symbol: trsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136747.4 | NP_610591.1 | 1 - 2 | 153 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

trsn| Gene Symbol: trsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136747.4 | NP_610591.1 | 1 - 2 | 153 | 115 | |
Set Membership: |
|