Assay Details
| Gene Symbol: trsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136747.4 | NP_610591.1 | 2 - 3 | 238 | 86 | |
| GenBank mRNA | AY118870.1 | - | 2 - 3 | 242 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

trsn| Gene Symbol: trsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136747.4 | NP_610591.1 | 2 - 3 | 238 | 86 | |
| GenBank mRNA | AY118870.1 | - | 2 - 3 | 242 | 86 | |
Set Membership: |
|