Assay Details
| Gene Symbol: trsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136747.4 | NP_610591.1 | 3 - 4 | 330 | 77 | |
| GenBank mRNA | AY118870.1 | - | 3 - 4 | 334 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

trsn| Gene Symbol: trsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136747.4 | NP_610591.1 | 3 - 4 | 330 | 77 | |
| GenBank mRNA | AY118870.1 | - | 3 - 4 | 334 | 77 | |
Set Membership: |
|