Assay Details
| Gene Symbol: metro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144170.2 | NP_001137642.1 | 4 - 5 | 884 | 76 | |
| NM_136798.3 | NP_610642.2 | 4 - 5 | 884 | 76 | ||
| GenBank mRNA | AF495381.1 | - | 4 - 5 | 853 | 76 | |
| BT003617.1 | - | 4 - 5 | 853 | 76 | ||
| BT003746.1 | - | 4 - 5 | 870 | 76 | ||
More Information
Set Membership: |
|

metro
Genomic Map