Assay Details
| Gene Symbol: metro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144170.2 | NP_001137642.1 | 6 - 7 | 1080 | 66 | |
| NM_001144171.2 | NP_001137643.1 | 5 - 6 | 1050 | 66 | ||
| NM_001299380.1 | NP_001286309.1 | 5 - 6 | 1050 | 66 | ||
| NM_136798.3 | NP_610642.2 | 6 - 7 | 1080 | 66 | ||
| GenBank mRNA | AF495381.1 | - | 6 - 7 | 1049 | 66 | |
| BT003617.1 | - | 6 - 7 | 1049 | 66 | ||
| BT003746.1 | - | 6 - 7 | 1066 | 66 | ||
More Information
Set Membership: |
|

metro
Genomic Map