Assay Details
| Gene Symbol: metro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144170.2 | NP_001137642.1 | 7 - 8 | 1400 | 60 | |
| NM_001299380.1 | NP_001286309.1 | 6 - 7 | 1370 | 60 | ||
| GenBank mRNA | BT003617.1 | - | 7 - 8 | 1369 | 60 | |
| BT003746.1 | - | 7 - 8 | 1386 | 60 | ||
More Information
Set Membership: |
|

metro
Genomic Map