Assay Details
| Gene Symbol: Cpr47Ea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136810.2 | NP_610654.1 | 2 - 3 | 223 | 80 | |
| GenBank mRNA | KX531261.1 | - | 1 - 2 | 178 | 80 | |
More Information
Set Membership: |
|

Cpr47Ea
Genomic Map