Assay Details
| Gene Symbol: Cpr47Ea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136810.2 | NP_610654.1 | 3 - 4 | 297 | 60 | |
| GenBank mRNA | KX531261.1 | - | 2 - 3 | 252 | 60 | |
More Information
Set Membership: |
|

Cpr47Ea
Genomic Map