Assay Details
| Gene Symbol: Cpr47Ee | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136815.2 | NP_610659.1 | 1 - 2 | 639 | 84 | |
| GenBank mRNA | BT100167.1 | - | 1 - 2 | 640 | 84 | |
More Information
Set Membership: |
|

Cpr47Ee
Genomic Map