Assay Details
| Gene Symbol: Cpr47Ef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136816.3 | NP_610660.2 | 11 - 12 | 1573 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpr47Ef| Gene Symbol: Cpr47Ef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136816.3 | NP_610660.2 | 11 - 12 | 1573 | 91 | |
Set Membership: |
|