Assay Details
| Gene Symbol: Cpr47Ef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201976.2 | NP_001188905.1 | 14 - 15 | 1734 | 101 | |
| NM_136816.3 | NP_610660.2 | 15 - 16 | 1767 | 101 | ||
More Information
Set Membership: |
|

Cpr47Ef
Genomic Map