Assay Details
| Gene Symbol: Cpr47Ef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201976.2 | NP_001188905.1 | 2 - 3 | 920 | 79 | |
| NM_136816.3 | NP_610660.2 | 2 - 3 | 920 | 79 | ||
More Information
Set Membership: |
|

Cpr47Ef
Genomic Map