Assay Details
| Gene Symbol: Smyd4-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136886.3 | NP_610730.1 | 2 - 3 | 1709 | 66 | |
| GenBank mRNA | AF145692.1 | - | 2 - 3 | 1685 | 66 | |
More Information
Set Membership: |
|

Smyd4-4
Genomic Map