Assay Details
| Gene Symbol: Cyp6g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299406.1 | NP_001286335.1 | 2 - 3 | 673 | 121 | |
| NM_136899.4 | NP_610743.2 | 2 - 3 | 660 | 121 | ||
| GenBank mRNA | BT050575.1 | - | 2 - 3 | 650 | 121 | |
More Information
Set Membership: |
|

Cyp6g1
Genomic Map