Assay Details
| Gene Symbol: Cyp6g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299406.1 | NP_001286335.1 | 3 - 4 | 1474 | 81 | |
| NM_136899.4 | NP_610743.2 | 3 - 4 | 1461 | 81 | ||
| GenBank mRNA | AY081960.1 | - | 2 - 3 | 1269 | 81 | |
| BT050575.1 | - | 3 - 4 | 1451 | 81 | ||
More Information
Set Membership: |
|

Cyp6g1
Genomic Map