Assay Details
| Gene Symbol: CG42700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299410.1 | NP_001286339.1 | 2 - 3 | 1142 | 98 | |
| NM_136910.3 | NP_610754.1 | 3 - 4 | 1123 | 98 | ||
More Information
Set Membership: |
|

CG42700
Genomic Map