Assay Details
| Gene Symbol: CG42700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201980.2 | NP_001188909.1 | 3 - 4 | 1103 | 73 | |
| NM_001273986.2 | NP_001260915.1 | 3 - 4 | 608 | 73 | ||
| NM_001273987.1 | NP_001260916.1 | 4 - 5 | 953 | 73 | ||
| NM_001273988.1 | NP_001260917.1 | 5 - 6 | 1115 | 73 | ||
| NM_001299409.1 | NP_001286338.1 | 3 - 4 | 1103 | 73 | ||
| NM_001299410.1 | NP_001286339.1 | 4 - 5 | 1520 | 73 | ||
| NM_001299411.1 | NP_001286340.1 | 3 - 4 | 1103 | 73 | ||
| NM_136910.3 | NP_610754.1 | 5 - 6 | 1501 | 73 | ||
| GenBank mRNA | BT088920.1 | - | 3 - 4 | 1101 | 73 | |
More Information
Set Membership: |
|

CG42700
Genomic Map