Assay Details
Gene Symbol: CG42700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201980.2 | NP_001188909.1 | 4 - 5 | 1214 | 72 | |
NM_001273986.2 | NP_001260915.1 | 4 - 5 | 719 | 72 | ||
NM_001273987.1 | NP_001260916.1 | 5 - 6 | 1064 | 72 | ||
NM_001273988.1 | NP_001260917.1 | 6 - 7 | 1226 | 72 | ||
NM_001299409.1 | NP_001286338.1 | 4 - 5 | 1214 | 72 | ||
NM_001299410.1 | NP_001286339.1 | 5 - 6 | 1631 | 72 | ||
NM_001299411.1 | NP_001286340.1 | 4 - 5 | 1214 | 72 | ||
NM_136910.3 | NP_610754.1 | 6 - 7 | 1612 | 72 | ||
GenBank mRNA | BT088920.1 | - | 4 - 5 | 1212 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
