Assay Details
| Gene Symbol: garz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136917.4 | NP_610761.2 | 9 - 10 | 5567 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

garz| Gene Symbol: garz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136917.4 | NP_610761.2 | 9 - 10 | 5567 | 76 | |
Set Membership: |
|