Assay Details
| Gene Symbol: Ak6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136953.4 | NP_610797.1 | 1 - 2 | 167 | 80 | |
| GenBank mRNA | AY071739.1 | - | 1 - 2 | 188 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ak6| Gene Symbol: Ak6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136953.4 | NP_610797.1 | 1 - 2 | 167 | 80 | |
| GenBank mRNA | AY071739.1 | - | 1 - 2 | 188 | 80 | |
Set Membership: |
|