Assay Details
| Gene Symbol: ClC-b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136954.4 | NP_610798.1 | 8 - 9 | 2389 | 127 | |
| GenBank mRNA | AY071581.1 | - | 8 - 9 | 2345 | 127 | |
More Information
Set Membership: |
|

ClC-b
Genomic Map