Assay Details
| Gene Symbol: CG17760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043074.2 | NP_001036539.1 | 6 - 7 | 1086 | 110 | |
| GenBank mRNA | AY122268.1 | - | 6 - 7 | 1061 | 110 | |
More Information
Set Membership: |
|

CG17760
Genomic Map