Assay Details
| Gene Symbol: CG17760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043074.2 | NP_001036539.1 | 7 - 8 | 1247 | 62 | |
| GenBank mRNA | AY122268.1 | - | 7 - 8 | 1222 | 62 | |
More Information
Set Membership: |
|

CG17760
Genomic Map