Assay Details
| Gene Symbol: SERCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299867.1 | NP_001286796.1 | 4 - 5 | 3049 | 112 | |
| NM_057484.3 | NP_476832.1 | 4 - 5 | 3049 | 112 | ||
| NM_166633.2 | NP_726381.1 | 5 - 6 | 2595 | 112 | ||
| NM_166634.2 | NP_726382.1 | 5 - 6 | 2605 | 112 | ||
| NM_166635.2 | NP_726383.1 | 5 - 6 | 2469 | 112 | ||
| NM_166636.2 | NP_726384.1 | 5 - 6 | 2520 | 112 | ||
| NM_166637.2 | NP_726385.1 | 4 - 5 | 3049 | 112 | ||
| NM_166638.2 | NP_726386.1 | 5 - 6 | 2479 | 112 | ||
| NM_166639.2 | NP_726387.1 | 5 - 6 | 2605 | 112 | ||
| GenBank mRNA | BT125984.1 | - | 5 - 6 | 2564 | 112 | |
More Information
Set Membership: |
|

SERCA
Genomic Map