Assay Details
| Gene Symbol: Galphas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169812.2 | NP_001163283.1 | 5 - 6 | 922 | 152 | |
| NM_001299869.1 | NP_001286798.1 | 4 - 5 | 1111 | 152 | ||
| NM_058157.5 | NP_477505.1 | 5 - 6 | 1135 | 152 | ||
| NM_058158.5 | NP_477506.1 | 5 - 6 | 1135 | 152 | ||
| NM_166640.3 | NP_726388.1 | 5 - 6 | 922 | 152 | ||
| GenBank mRNA | AY058572.1 | - | 5 - 6 | 906 | 152 | |
| M23233.1 | - | 5 - 6 | 922 | 152 | ||
More Information
Set Membership: |
|

Galphas
Genomic Map