Assay Details
| Gene Symbol: Not11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299884.1 | NP_001286813.1 | 1 - 2 | 519 | 85 | |
| NM_138030.4 | NP_611874.1 | 1 - 2 | 312 | 85 | ||
| NM_166652.4 | NP_726406.1 | 1 - 2 | 312 | 85 | ||
| GenBank mRNA | BT024271.1 | - | 1 - 2 | 403 | 85 | |
More Information
Set Membership: |
|

Not11
Genomic Map