Assay Details
| Gene Symbol: Eps-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259573.1 | NP_001246502.1 | 3 - 4 | 1363 | 56 | |
| NM_138121.2 | NP_611965.2 | 3 - 4 | 1363 | 56 | ||
| NM_166686.1 | NP_726481.1 | 3 - 4 | 1363 | 56 | ||
| NM_166687.1 | NP_726482.1 | 3 - 4 | 1363 | 56 | ||
| GenBank mRNA | BT125713.1 | - | 3 - 4 | 1329 | 56 | |
More Information
Set Membership: |
|

Eps-15
Genomic Map