Assay Details
| Gene Symbol: Dll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079133.2 | NP_523857.1 | 4 - 5 | 1262 | 66 | |
| NM_166689.2 | NP_726486.1 | 4 - 5 | 1262 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dll| Gene Symbol: Dll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079133.2 | NP_523857.1 | 4 - 5 | 1262 | 66 | |
| NM_166689.2 | NP_726486.1 | 4 - 5 | 1262 | 66 | ||
Set Membership: |
|