Assay Details
| Gene Symbol: Dll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144287.2 | NP_001137759.1 | 4 - 5 | 1440 | 48 | |
| NM_079133.2 | NP_523857.1 | 5 - 6 | 1365 | 48 | ||
| NM_166689.2 | NP_726486.1 | 5 - 6 | 1365 | 48 | ||
| GenBank mRNA | BT030126.1 | - | 4 - 5 | 1423 | 48 | |
More Information
Set Membership: |
|

Dll
Genomic Map