Assay Details
| Gene Symbol: CG44247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138130.4 | NP_611974.2 | 2 - 3 | 1563 | 64 | |
| NM_166690.3 | NP_726489.2 | 2 - 3 | 1563 | 64 | ||
| GenBank mRNA | BT100206.1 | - | 2 - 3 | 1520 | 64 | |
More Information
Set Membership: |
|

CG44247
Genomic Map