Assay Details
| Gene Symbol: pan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258477.2 | NP_001245406.1 | 10 - 11 | 1539 | 105 | |
| NM_079880.6 | NP_524619.3 | 11 - 12 | 1666 | 105 | ||
| NM_166718.3 | NP_726522.1 | 10 - 11 | 1407 | 105 | ||
| NM_166720.3 | NP_726524.1 | 10 - 11 | 1373 | 105 | ||
| NM_166723.3 | NP_726527.2 | 11 - 12 | 1798 | 105 | ||
| NM_166724.4 | NP_726528.2 | 3 - 4 | 3283 | 105 | ||
| GenBank mRNA | BT044547.1 | - | 12 - 13 | 1303 | 105 | |
| BT053747.1 | - | 10 - 11 | 1275 | 105 | ||
| U88538.1 | - | 10 - 11 | 1332 | 105 | ||
| Y09125.1 | - | 1514 | 105 | |||
More Information
Set Membership: |
|

pan
Genomic Map