Assay Details
| Gene Symbol: pan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014685.3 | NP_001014685.1 | 4 - 5 | 902 | 131 | |
| NM_001038709.3 | NP_001033798.1 | 4 - 5 | 902 | 131 | ||
| NM_001258476.2 | NP_001245405.1 | 4 - 5 | 902 | 131 | ||
| NM_001258477.2 | NP_001245406.1 | 3 - 4 | 643 | 131 | ||
| NM_079880.6 | NP_524619.3 | 4 - 5 | 770 | 131 | ||
| NM_166718.3 | NP_726522.1 | 3 - 4 | 511 | 131 | ||
| NM_166720.3 | NP_726524.1 | 3 - 4 | 477 | 131 | ||
| NM_166723.3 | NP_726527.2 | 4 - 5 | 902 | 131 | ||
| GenBank mRNA | BT021431.1 | - | 4 - 5 | 835 | 131 | |
| BT044547.1 | - | 5 - 6 | 407 | 131 | ||
| BT053747.1 | - | 3 - 4 | 379 | 131 | ||
| U88538.1 | - | 3 - 4 | 436 | 131 | ||
| Y09125.1 | - | 618 | 131 | |||
More Information
Set Membership: |
|

pan
Genomic Map