Assay Details
| Gene Symbol: pan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014685.3 | NP_001014685.1 | 8 - 9 | 1285 | 95 | |
| NM_001038709.3 | NP_001033798.1 | 8 - 9 | 1285 | 95 | ||
| NM_001258476.2 | NP_001245405.1 | 8 - 9 | 1285 | 95 | ||
| NM_001258477.2 | NP_001245406.1 | 7 - 8 | 1026 | 95 | ||
| NM_079880.6 | NP_524619.3 | 8 - 9 | 1153 | 95 | ||
| NM_166718.3 | NP_726522.1 | 7 - 8 | 894 | 95 | ||
| NM_166720.3 | NP_726524.1 | 7 - 8 | 860 | 95 | ||
| NM_166723.3 | NP_726527.2 | 8 - 9 | 1285 | 95 | ||
| GenBank mRNA | BT021431.1 | - | 8 - 9 | 1218 | 95 | |
| BT044547.1 | - | 9 - 10 | 790 | 95 | ||
| BT053747.1 | - | 7 - 8 | 762 | 95 | ||
| U88538.1 | - | 7 - 8 | 819 | 95 | ||
| Y09125.1 | - | 1001 | 95 | |||
More Information
Set Membership: |
|

pan
Genomic Map