Assay Details
| Gene Symbol: pan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258477.2 | NP_001245406.1 | 9 - 10 | 1366 | 83 | |
| NM_079880.6 | NP_524619.3 | 10 - 11 | 1493 | 83 | ||
| NM_166718.3 | NP_726522.1 | 9 - 10 | 1234 | 83 | ||
| NM_166720.3 | NP_726524.1 | 9 - 10 | 1200 | 83 | ||
| NM_166723.3 | NP_726527.2 | 10 - 11 | 1625 | 83 | ||
| NM_166724.4 | NP_726528.2 | 2 - 3 | 3110 | 83 | ||
| GenBank mRNA | BT044547.1 | - | 11 - 12 | 1130 | 83 | |
| BT053747.1 | - | 9 - 10 | 1102 | 83 | ||
| U88538.1 | - | 9 - 10 | 1159 | 83 | ||
| Y09125.1 | - | 1341 | 83 | |||
More Information
Set Membership: |
|

pan
Genomic Map