Assay Details
| Gene Symbol: CaMKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079883.3 | NP_524622.1 | 2 - 3 | 1667 | 88 | |
| NM_166759.2 | NP_726569.1 | 3 - 4 | 524 | 88 | ||
| NM_166760.2 | NP_726570.1 | 4 - 5 | 865 | 88 | ||
| GenBank mRNA | BT001637.1 | - | 4 - 5 | 475 | 88 | |
| Y17917.1 | - | 1608 | 88 | |||
More Information
Set Membership: |
|

CaMKI
Genomic Map