Assay Details
| Gene Symbol: CaMKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079883.3 | NP_524622.1 | 3 - 4 | 1709 | 118 | |
| NM_166759.2 | NP_726569.1 | 4 - 5 | 566 | 118 | ||
| NM_166760.2 | NP_726570.1 | 5 - 6 | 907 | 118 | ||
| GenBank mRNA | BT001637.1 | - | 5 - 6 | 517 | 118 | |
| Y17917.1 | - | 1650 | 118 | |||
More Information
Set Membership: |
|

CaMKI
Genomic Map