Assay Details
| Gene Symbol: CaMKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079883.3 | NP_524622.1 | 6 - 7 | 2187 | 106 | |
| NM_166759.2 | NP_726569.1 | 7 - 8 | 1044 | 106 | ||
| NM_166760.2 | NP_726570.1 | 8 - 9 | 1385 | 106 | ||
| NM_166761.2 | NP_726571.2 | 6 - 7 | 1409 | 106 | ||
| NM_166764.2 | NP_726574.1 | 5 - 6 | 755 | 106 | ||
| GenBank mRNA | BT001637.1 | - | 8 - 9 | 995 | 106 | |
| Y17917.1 | - | 2128 | 106 | |||
More Information
Set Membership: |
|

CaMKI
Genomic Map