Assay Details
| Gene Symbol: Asator | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272126.2 | NP_001259055.1 | 13 - 14 | 4031 | 91 | |
| NM_001272127.1 | NP_001259056.1 | 13 - 14 | 4618 | 91 | ||
| NM_143667.3 | NP_651924.2 | 13 - 14 | 3954 | 91 | ||
| NM_166766.3 | NP_726577.2 | 12 - 13 | 4288 | 91 | ||
More Information
Set Membership: |
|

Asator
Genomic Map