Assay Details
| Gene Symbol: Asator | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272125.1 | NP_001259054.1 | 2 - 3 | 654 | 60 | |
| NM_001272126.2 | NP_001259055.1 | 2 - 3 | 731 | 60 | ||
| NM_001272127.1 | NP_001259056.1 | 2 - 3 | 1318 | 60 | ||
| NM_001272128.1 | NP_001259057.1 | 2 - 3 | 1318 | 60 | ||
| NM_001272129.1 | NP_001259058.1 | 2 - 3 | 797 | 60 | ||
| NM_001297785.1 | NP_001284714.1 | 2 - 3 | 641 | 60 | ||
| NM_143667.3 | NP_651924.2 | 2 - 3 | 654 | 60 | ||
| NM_166765.3 | NP_726576.2 | 2 - 3 | 654 | 60 | ||
| NM_166766.3 | NP_726577.2 | 2 - 3 | 797 | 60 | ||
| GenBank mRNA | BT133187.1 | - | 2 - 3 | 620 | 60 | |
More Information
Set Membership: |
|

Asator
Genomic Map