Assay Details
| Gene Symbol: Pur-alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272132.1 | NP_001259061.1 | 6 - 7 | 835 | 74 | |
| NM_001272133.1 | NP_001259062.1 | 6 - 7 | 652 | 74 | ||
| NM_001272134.1 | NP_001259063.1 | 6 - 7 | 660 | 74 | ||
| NM_079885.2 | NP_524624.1 | 6 - 7 | 758 | 74 | ||
| NM_166767.2 | NP_726579.1 | 6 - 7 | 755 | 74 | ||
| NM_166768.2 | NP_726580.1 | 6 - 7 | 680 | 74 | ||
| NM_166769.2 | NP_726581.1 | 6 - 7 | 660 | 74 | ||
| GenBank mRNA | AY061185.1 | - | 6 - 7 | 727 | 74 | |
| BT126365.1 | - | 6 - 7 | 660 | 74 | ||
More Information
Set Membership: |
|

Pur-alpha
Genomic Map