Assay Details
| Gene Symbol: ey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014693.2 | NP_001014693.1 | 4 - 5 | 1157 | 104 | |
| NM_001014694.2 | NP_001014694.1 | 3 - 4 | 597 | 104 | ||
| NM_079889.3 | NP_524628.2 | 3 - 4 | 977 | 104 | ||
| NM_166789.2 | NP_726607.1 | 3 - 4 | 549 | 104 | ||
| GenBank mRNA | BT011390.1 | - | 3 - 4 | 549 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT011390.1, NM_166789.2 transcripts.
Set Membership: |
|

ey
Genomic Map