Assay Details
| Gene Symbol: ey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014693.2 | NP_001014693.1 | 5 - 6 | 1805 | 70 | |
| NM_001014694.2 | NP_001014694.1 | 4 - 5 | 1245 | 70 | ||
| NM_079889.3 | NP_524628.2 | 4 - 5 | 1625 | 70 | ||
| NM_166789.2 | NP_726607.1 | 4 - 5 | 1197 | 70 | ||
| GenBank mRNA | BT011390.1 | - | 4 - 5 | 1197 | 70 | |
More Information
Set Membership: |
|

ey
Genomic Map