Assay Details
| Gene Symbol: ey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014693.2 | NP_001014693.1 | 7 - 8 | 2252 | 74 | |
| NM_001014694.2 | NP_001014694.1 | 6 - 7 | 1692 | 74 | ||
| NM_079889.3 | NP_524628.2 | 6 - 7 | 2072 | 74 | ||
| NM_166789.2 | NP_726607.1 | 6 - 7 | 1644 | 74 | ||
| GenBank mRNA | BT011390.1 | - | 6 - 7 | 1644 | 74 | |
More Information
Set Membership: |
|

ey
Genomic Map