Assay Details
| Gene Symbol: ey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014693.2 | NP_001014693.1 | 8 - 9 | 2609 | 63 | |
| NM_001014694.2 | NP_001014694.1 | 7 - 8 | 2049 | 63 | ||
| NM_079889.3 | NP_524628.2 | 7 - 8 | 2429 | 63 | ||
| NM_166789.2 | NP_726607.1 | 7 - 8 | 2001 | 63 | ||
| GenBank mRNA | BT011390.1 | - | 7 - 8 | 2001 | 63 | |
More Information
Set Membership: |
|

ey
Genomic Map